Learn More
MAP2 Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
Brand:  Novus Biologicals NB300-213
Product Code. 18748383
  • 516.00€ / 0.05mL
Explore more special offers
Description
Description
MAP2 Polyclonal antibody specifically detects MAP2 in Human, Mouse, Rat, Feline samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Knockdown.
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
MAP2 Antibody, Novus Biologicals™ > 0.05mL; Unlabeled