Learn More
Plum™ Plutect Dual Skin Cream
Skin care cream for use before and during work with alternating work processes involving water-based and other substances. May also be applied as replenishing after-work skin cream.
Brand:  Plum™ P2503
 View more versions of this product
Product Code. 15721137
  • 275.00€ / Pack of 8
Estimated Shipment: 08-05-2025
to see stock.
Explore more special offers
Description
  • Effective protection relative to water-based and other substances
  • Perfume-, colour- and paraben-free
  • Resistant to 20 out of 20 substances in Suskind test/winner of BUS test 2007
  • Quickly absorbed, eases skin cleaning
  • Vaseline and zinc oxide provides skin protection and reduces skin irritation
  • Specifications
    Specifications
    Skin Care Cream
    Bag-in-box
    0.7 L
    MP2000 dispenser
    8 Pk.
    Product Title
    Plum™ Plutect Dual Skin Cream > Packaging: 8 x 0.7 litre bag-in-box