missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Pinacidil Monohydrate K+ channel activator

K+ channel activator

Brand:  Novus Biologicals™ NBP2-62756-10MG

 View more versions of this product

Product Code. 18227984

  • 64.00€ / 10mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

PubChem ID: 55329

Application Notes: May be dissolved in DMSO (25 mg/mL, warm); or ethanol (25 mg/mL, warm)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

85371-64-8
C13H19N5· H2O
10 mg
Pinacidil Monohydrate K+ channel activator
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
>98% (HPLC); NMR (conforms)
Synthetic
Powder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Novus Biologicals™ Pinacidil Monohydrate K+ channel activator > 10 mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.