missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Pinacidil Monohydrate K+ channel activator

Product Code. p-7146022 Shop All Bio Techne Products
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18227984

Brand: Novus Biologicals™ NBP26275610MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

K+ channel activator

PubChem ID: 55329

Application Notes: May be dissolved in DMSO (25 mg/mL, warm); or ethanol (25 mg/mL, warm)

TRUSTED_SUSTAINABILITY

Specifications

CAS 85371-64-8
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Molecular Formula C13H19N5· H2O
Purity >98% (HPLC); NMR (conforms)
Quantity 10 mg
Source Synthetic
Target Pinacidil Monohydrate K+ channel activator
Form Powder
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.