missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Glutathione Spin Columns, 3 mL

Purify GST-tagged recombinant fusion proteins from cell lysates using glutathione (GSH) beaded agarose affinity purification resins, spin columns and plates.

Brand:  Thermo Scientific™ 16105

480.70 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.


Product Code. 11862104

  • 514.00€ / Pack of 5

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

3 mL
Upon receipt store at 4°C.
Pierce™
Recombinant
Spin Column
Spin Column
Pierce Glutathione Spin Columns, 3mL
Liquid
5 Columns
GST-Tagged Protein
Glutathione
Affinity, Agarose Resin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM345750 Certificate TypeCertificate of Compliance Date04/12/2024 Catalog Number16105
Lot NumberM345740 Certificate TypeCertificate of Compliance Date04/12/2024 Catalog Number16105
Lot NumberM345730 Certificate TypeCertificate of Compliance Date04/12/2024 Catalog Number16105
Lot NumberM345720 Certificate TypeCertificate of Compliance Date04/12/2024 Catalog Number16105
Lot NumberZH403079 Certificate TypeCertificate of Analysis Date31/08/2024 Catalog Number16105
Promotions

Promotions

  • Multiple Manufacturers
    Advance your proteomics research with essential tools
    Promo Code: 25977

    Expires2025-05-31

    Days

    43

    Hours

    19

    Minutes

    43

    Seconds

    32
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Glutathione Spin Columns, 3 mL > Spin columns, 22mL-capacity, 3mL resin; 5 columns

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.