Learn More
DWK Life Sciences Kimble™ Phenolic Screw Cap with Liner
Secure your media during autoclaving with the Kimble™ Phenolic Screw Cap with Liner. It is made of a special phenolic material (wood filled) and very resistant to the effects of temperature and steam encountered in autoclaving
Brand:  DWK Life Sciences 73800-13415
 View more versions of this product
Product Code. 11868532
  • 283.00€ / Pack of 1000
Estimated Shipment: 07-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Secure your media during autoclaving with the Kimble™ Phenolic Screw Cap with Liner. It is made of a special phenolic material (wood filled) and very resistant to the effects of temperature and steam encountered in autoclaving
  • Styrene Liner
  • Screw Cap
TRUSTED_SUSTAINABILITY
Specifications
Specifications
13-415
Autoclavable
Phenolic
1000 Pack
Rubber
Fixed Screw Cap
Black
Centrifuge Tubes
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Promotions
Promotions
Product Content Correction
Product Title
DWK Life Sciences Kimble™ Phenolic Screw Cap with Liner > Black with white rubber liners