Learn More
DWK Life Sciences Kimble™ Phenolic Screw Cap with Liner
Secure your media during autoclaving with the Kimble™ Phenolic Screw Cap with Liner. It is made of a special phenolic material (wood filled) and very resistant to the effects of temperature and steam encountered in autoclaving
283.00€ - 421.00€
Specifications
Material Phenolic
View More Specs
Products 2
Product Code Brand Liner Material Price Quantity & Availability  
Product Code Brand Liner Material Price Quantity & Availability  
11868532
View Documents
DWK Life Sciences
73800-13415
Rubber
283.00€
Pack of 1000
Estimated Shipment: 11-06-2025
to see stock.
Add to basket
11883825
View Documents
DWK Life Sciences
73800-15415
Rubber
421.00€
Pack of 1000
In Stock
to see stock.
Edge
Add to basket
Description
Description
Secure your media during autoclaving with the Kimble™ Phenolic Screw Cap with Liner. It is made of a special phenolic material (wood filled) and very resistant to the effects of temperature and steam encountered in autoclaving
  • Styrene Liner
  • Screw Cap
Specifications
Specifications
Phenolic
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
DWK Life Sciences Kimble™ Phenolic Screw Cap with Liner