missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ ORP Test Solution

Manufactured from reagent grade chemicals and used to test platinum and gold ORP electrodes. Hanna Instruments™ ORP Test Solution includes lot number and expiration date clearly marked on the label to ensure product quality.

Brand:  Hanna Instruments™ HI7022L


Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 11534317

  • 46.15€ / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

ORP test solution made from reagent grade chemicals and used to test platinum and gold ORP electrodes.
  • Lot number and expiration date clearly marked on the label
  • Air tight with a tamper-proof seal to ensure the quality of the solution.
  • Prepared with reagent grade chemicals
  • Allows for a quick check of your ORP electrode
Specifications

Specifications

470mV
For Testing the Performance of ORP Electrodes
500 mL
ORP Test Solution
Air Tight Bottle With Tamper-Proof Seal
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hanna Instruments™ ORP Test Solution > Sample Size: 500mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.