missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ INSTA-Blot Mouse Brain

PVDF membrane containing denatured protein from cells or tissue lysates

Brand:  Novus Biologicals™ NBP2-30120

Product Code. 18175922

  • 551.00€ / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Screening proteins from various human, mouse and rat tissues
  • Contains mouse brain lysate (30μg total protein per lane) on 4-20% Tris-Glycine mini gel
  • Prepared from resolution by SDS-PAGE at 125V for 1 hour
  • Proteins are electro-blotted for 2 hours at 25V onto PVDF membrane
  • Stained with amido black for visualization of proteins

Recommended Storage

Store at room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Screening proteins from various human, mouse and rat tissues
1 Membrane
PVDF
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Novus Biologicals™ INSTA-Blot Mouse Brain >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.