missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Masterflex™ Nylon Male Threaded Adapter Barbed Fitting
Adapts from a NPT male thread to a hose barb for quick and easy fluid path assembly in the laboratory, without the use of tools.
Brand: Masterflex™ 31702-02
This item is not returnable.
View return policy
Description
- No leaks, flow restrictions or costly downtime
- A crucial component in setting up a fluid handling system
- Typically holds pressures up to 30 psi (2 bar)
- Slip flexible tubing or hose onto the barb to create a secure seal
Specifications
Nylon | |
10 Pack | |
Straight | |
Barbed Fitting |
1/8 x 3/16 in. tubing I.D. | |
Ethylene Oxide | |
30 psi (2 bar) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Masterflex™ Nylon Male Threaded Adapter Barbed Fitting > For 1/8 x 3/16 in. tubing I.D.
Spot an opportunity for improvement?Share a Content Correction