missing translation for 'onlineSavingsMsg'
Learn More

NICE-3 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-97389PE

Product Code. 18685792

  • 407.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

NICE-3 Polyclonal antibody specifically detects NICE-3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NICE-3
Polyclonal
Immunohistochemistry-Paraffin
chromosome 1 open reading frame 43, HCV NS5A-transactivated protein 4
Produced in rabbits immunized with E. coli-derived Human NICE-3 fragment.
0.1 mL
Primary
Human
Purified
Immunohistochemistry (Paraffin)
PE
PBS
Rabbit
Antigen and protein A Affinity-purified
RUO
25912
Store at 4C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
NICE-3 Antibody, PE, Novus Biologicals™ > 0.1 mL; PE

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.