missing translation for 'onlineSavingsMsg'
Learn More

Wheaton™ Bench-Top Mini-Bottle Roller Apparatus

For cell cultivation research or staining procedures. Wheaton™ Bench-Top Mini-Bottle Roller Apparatus allows for serum and media bottles, as well as culture tubes.

Brand:  Wheaton™ W348923-C

 View more versions of this product

Product Code. 10185812

  • 2500.00€ / Each
Estimated Shipment: 30-04-2025
to see stock.
Explore more special offers
Add to basket

Récupel-bijdrage uitgesloten
This item is not returnable. View return policy

Description

Description

  • Accommodates bottles 38 to 60mm in diameter and 240mm long
  • Deck kits available to double or triple capacity
  • Precise and reproducible speed adjustments
TRUSTED_SUSTAINABILITY
Specifications

Specifications

3 to 45 rpm
100/240 V
14 W
50/60 Hz
18 cm
8 kg
For the growth and observation of various cell cultures
100/240 V, 50/60 Hz, 14 W
40°C
CE, CSA, UL
32.5 cm
32 cm
C Model
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
View accessory products
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Wheaton™ Bench-Top Mini-Bottle Roller Apparatus > Model: C Model

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.