Learn More
Neuroligin 3/NLGN3 Antibody (242), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-89824H
Product Code. 18672961
  • 612.00€ / 0.10mL
Explore more special offers
Description
Description
Neuroligin 3/NLGN3 Monoclonal antibody specifically detects Neuroligin 3/NLGN3 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Neuroligin 3/NLGN3
Monoclonal
HRP
PBS
Rabbit
Protein A purified
RUO
Primary
Human
Purified
Flow Cytometry
242
Flow Cytometry
ASPGX1, AUTSX1, Gliotactin homolog, KIAA1480HNL3, neuroligin 3, neuroligin-3, NL3
This antibody was obtained from a rabbit immunized with purified, recombinant Human Neuroligin 3/NLGN3 (Uniprot#: Q9NZ94-2; Met1-Ser689).
0.1 mL
Neuroscience
54413
Store at 4C in the dark.
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Neuroligin 3/NLGN3 Antibody (242), HRP, Novus Biologicals™ > 0.1 mL; HRP