Learn More
Genevac™ miVac Centrifugal Concentrators
Choose the ideal concentrator according to workload. Genevac™ miVac Centrifugal Concentrators are modular systems for removing water and solvents from samples. They are available with a range of unique JetRotors™ and vacuum pumps. An optional high-power cold trap is used to condense solvent vapors.
Brand:  Genevac™ QUC-23050-B00
 View more versions of this product
Product Code. 10353893
  • 8845.35€ / Each
Explore more special offers
Récupel-bijdrage uitgesloten
Description
Show More
Specifications
Specifications
miVac Quatro
Samples in tubes, microplates and vials
Concentrator with large concentration chamber
480W x 594D x 300H mm
230 V 50/60 Hz 650 w
30°C to 80°C
200 Reaction Vessels (0.5 mL), 20 Microtiter Plates
Max G-force: 250
35 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Manuals
Product Information
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
Genevac™ miVac Centrifugal Concentrators > Model: miVac Quattro; Capacity: 200 reaction vessels 0.5mL or 20 microtiter plates; Includes concentrator with large chamber