Learn More
Fisherbrand™ Natural Rubber Two Hole Stoppers
Used for laboratory purposes.
Brand:  Fisherbrand™ RB037/2H
 View more versions of this product
Product Code. 11587632
  • 31.00€ / Pack of 10
Estimated Shipment: 28-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Used for laboratory purposes.
  • Made up of natural rubber
  • Red in color
  • Stoppers with two holes
Specifications
Specifications
5 mm
46/55 IRHD
10/Pk.
37 mm
Natural Rubber
For laboratory use
38.5 mm
Red
42.5 mm
Tapered Stopper (2 holes)
Product Title
Fisherbrand™ Natural Rubber Two Hole Stoppers > Bottom Diameter: 37mm; Top Diameter: 42.5mm