Learn More
BINDER™ Swivel Casters
Height-adjustable swivel casters
Brand: BINDER™ 8012-2541
This item is not returnable.
View return policy
Description
Height-adjustable swivel casters for horizontal alignment of unit on uneven floors.
Specifications
KB 260, KB 470, KB 720, KB 1060 | |
Steel | |
Castor Frame |
Product Content Correction
Product Title
BINDER™ Swivel Casters > For use with KB 260, KB 470, KB 720, KB 1060