Learn More
R&D Systems™ Mouse CCL17/TARC Quantikine ELISA Kit
Mouse CCL17/TARC Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand: R&D Systems™ MCC170
Description
The Mouse CCL17/TARC Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse CCL17/TARC in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul).
Specifications
7.80 - 500pg/mL | |
HRP | |
Natural and Recombinant Mouse TARC | |
20295 | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |
5pg/mL | |
Cell Culture Supernatant, Plasma (EDTA), Serum | |
ELISA | |
Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL) | |
ABCD-2, CC chemokine TARC, C-C motif chemokine 17, chemokine (C-C motif) ligand 17, MGC138273, SCYA17, SCYA17MGC138271, small inducible cytokine subfamily A (Cys-Cys), member 17, Small-inducible cytokine A17, T cell-directed CC chemokine, TARC, TARCA-152E5.3, Thymus and activation-regulated chemokine |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.