Learn More
Thermo Scientific™ Mixtainer BioProcess Container (BPC) for HyPerforma™ Mixtainer™ Systems
The complete Thermo Scientific™ Mixtainer™ System is an integrated, single-use sterile unit for mixing of cell culture media, and other process liquids such as buffers, reagents and bulk drug precursors or products.
1735.00€ - 10325.00€
Specifications
Tubing Style TPE
View More Specs
Products 4
Product Code Brand Capacity (Metric) Port Detail Price Quantity & Availability  
Product Code Brand Capacity (Metric) Port Detail Price Quantity & Availability  
30040271
View Documents
Thermo Scientific™
SH30661.01
50 L 4 x Top-mounted Ports and Tubing (2 with 0.25 in. ID Tubing and male/female Luer connectors, 2 with 3/1 in. ID tubing and quick-connect couplings)
9280.00€
Pack of 15
30040272
View Documents
Thermo Scientific™
SH30661.02
100 L 4 x Top-mounted Ports and Tubing (2 with 0.25 in. ID Tubing and male/female Luer connectors, 2 with 3/1 in. ID tubing and quick-connect couplings)
10325.00€
Pack of 15
30040270
View Documents
Thermo Scientific™
SH30661.03
200 L 4 x Top-mounted Ports and Tubing (2 with 0.25 in. ID Tubing and male/female Luer connectors, 2 with 3/1 in. ID tubing and quick-connect couplings)
4320.00€
Pack of 5
30040273
View Documents
Thermo Scientific™
SH30687.01
50 L Powder Port
1735.00€
Pack of 3
Description
Description
Recommended for:
  • Hydration of powdered media and buffers
  • Sterile mixing of cell culture media with sera and reagents
  • Sterile mixing of buffer solutions or other process fluids
  • Pooling of sterile liquid fractions
Specifications
Specifications
TPE
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Mixtainer BioProcess Container (BPC) for HyPerforma™ Mixtainer™ Systems