Learn More
Thermo Scientific™ Mixtainer BioProcess Container (BPC) for HyPerforma™ Mixtainer™ Systems
The complete Thermo Scientific™ Mixtainer™ System is an integrated, single-use sterile unit for mixing of cell culture media, and other process liquids such as buffers, reagents and bulk drug precursors or products.
Brand: Thermo Scientific™ SH30661.03
This item is not returnable.
View return policy
Description
Recommended for:- Hydration of powdered media and buffers
- Sterile mixing of cell culture media with sera and reagents
- Sterile mixing of buffer solutions or other process fluids
- Pooling of sterile liquid fractions
Specifications
200 L | |
4 x Top-mounted Ports and Tubing (2 with 0.25 in. ID Tubing and male/female Luer connectors, 2 with 3/1 in. ID tubing and quick-connect couplings) | |
TPE | |
Sterile |