missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ MDH1 Protein

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

403.00€ - 1025.00€

Specifications

Concentration 1.0mg/mL
For Use With (Application) ELISA, SDS-PAGE
Formulation In 20mM Tris-HCl Buffer (pH 8.0) containing 10% Glycerol
Gene ID (Entrez) 4190
Molecular Weight (g/mol) 37.4kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18273155
View Documents
Novus Biologicals™
NBP1-45315-0.1MG
0.1 mg
403.00€
0.10mg
Please to purchase this item. Need a web account? Register with us today!
18295344
View Documents
Novus Biologicals™
NBP1-45315-0.5MG
0.5 mg
1025.00€
0.50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

A bioactive recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-334 of Human MDH1 The Recombinant Human MDH1 Protein is derived from E. coli. The Recombinant Human MDH1 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
Specifications

Specifications

1.0mg/mL
In 20mM Tris-HCl Buffer (pH 8.0) containing 10% Glycerol
37.4kDa
Human
Store at -80°C. Avoid freeze-thaw cycles.
MDH1
Recombinant
ELISA, SDS-PAGE
4190
Protein
MSEPIRVLVT GAAGQIAYSL LYSIGNGSVF GKDQPIILVL LDITPMMGVL DGVLMELQDC ALPLLKDVIATDKEDVAFKD LDVAILVGSM PRREGMERKD LLKANVKIFK SQGAALDKYA KKSVKVIVVG NPANTNCLTA SKSAPSIPKE NFSCLTRLDH NRAKAQIALK LGVTANDVKN VIIWGNHSST QYPDVNHAKV KLQGKEVGVY EALKDDSWLK GEFVTTVQQR GAAVIKARKL SSAMSAAKAI CDHVRDIWFG TPEGEFVSMG VISDGNSYGV PDDLLYSFPV VIKNKTWKFV EGLPINDFSR EKMDLTAKEL TEEKESAFEF LSSALEHHHH HH
RUO
Unconjugated
>95%
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Novus Biologicals™ MDH1 Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.