missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MDH1 Protein
Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity
Brand: Novus Biologicals™ NBP1-45315-0.1MG
This item is not returnable.
View return policy
Description
A bioactive recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-334 of Human MDH1 The Recombinant Human MDH1 Protein is derived from E. coli. The Recombinant Human MDH1 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
Specifications
1.0mg/mL | |
In 20mM Tris-HCl Buffer (pH 8.0) containing 10% Glycerol | |
37.4kDa | |
0.1 mg | |
MSEPIRVLVT GAAGQIAYSL LYSIGNGSVF GKDQPIILVL LDITPMMGVL DGVLMELQDC ALPLLKDVIATDKEDVAFKD LDVAILVGSM PRREGMERKD LLKANVKIFK SQGAALDKYA KKSVKVIVVG NPANTNCLTA SKSAPSIPKE NFSCLTRLDH NRAKAQIALK LGVTANDVKN VIIWGNHSST QYPDVNHAKV KLQGKEVGVY EALKDDSWLK GEFVTTVQQR GAAVIKARKL SSAMSAAKAI CDHVRDIWFG TPEGEFVSMG VISDGNSYGV PDDLLYSFPV VIKNKTWKFV EGLPINDFSR EKMDLTAKEL TEEKESAFEF LSSALEHHHH HH | |
RUO | |
Unconjugated | |
>95% |
ELISA, SDS-PAGE | |
4190 | |
Protein | |
Human | |
Store at -80°C. Avoid freeze-thaw cycles. | |
MDH1 | |
Recombinant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ MDH1 Protein > 0.1mg
Spot an opportunity for improvement?Share a Content Correction