missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ L-755,507
Very potent and selective β3 partial agonist
253.00€ - 987.00€
Specifications
Purity | >98% |
---|---|
Product Type | L-755,507 |
Molecular Weight (g/mol) | 584.73 |
Inhibitors | L-755,507 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18790878
|
Tocris Bioscience™
2197/10 |
10 mg |
253.00€
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18075602
|
Tocris Bioscience™
2197/50 |
50 mg |
987.00€
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent β3-adrenergic receptor partial agonist > 1000-fold selective over β1- and β2-adrenoceptors (EC50 values are 0.43, 580 and > 10000 nM for activation of cloned human β3-, β1- and β2-adrenoceptors respectively). Stimulates lipolysis in rhesus adipocytes in vitro (EC50 = 3.9 nM). Enhances CRISPR-mediated homology-directed repair (HDR) efficiency 2-3-fold for large fragments and ∽9-fold for point mutations, in human induced pluripotent stem cells (iPSCs).Specifications
>98% | |
L-755,507 | |
584.73 | |
L-755,507 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title