Learn More
Tocris Bioscience™ L-755,507
Very potent and selective β3 partial agonist
Brand:  Tocris Bioscience™ 2197/10
 View more versions of this product
Product Code. 18790878
  • 253.00€ / 10mg
Explore more special offers
Description
Description
L-755,507 is a potent β3-adrenergic receptor partial agonist > 1000-fold selective over β1- and β2-adrenoceptors (EC50 values are 0.43, 580 and > 10000 nM for activation of cloned human β3-, β1- and β2-adrenoceptors respectively). Stimulates lipolysis in rhesus adipocytes in vitro (EC50 = 3.9 nM). Enhances CRISPR-mediated homology-directed repair (HDR) efficiency 2-3-fold for large fragments and ∽9-fold for point mutations, in human induced pluripotent stem cells (iPSCs).
Specifications
Specifications
>98%
L-755,507
L-755,507
10 mg
584.73