Learn More
Mitutoyo™ IP54 Digital Micrometer
Highly robust digital micrometer with outstanding accuracy. Mitutoyo™ IP54 Digital Micrometer features extraordinary battery life.
Brand: Mitutoyo™ 293240
Description
- Outstanding accuracy.
- Oil-resistant material used for all plastic parts.
- Extraordinary battery life of 2,4 years.
- Excellent resistance against water and dust (IP65 protection level) enables this micrometer to be used in machining situations that include splashing coolant fluid.
Specifications
Micrometer | |
Digital | |
0 to 25 mm |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.