Learn More
BINDER™ Interior Lighting
Interior lighting with climate-resistant LEDs.
903.00€ - 1608.00€
Specifications
Material Glass
Type Lighting
View More Specs
Products 13
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
30238222
View Documents
BINDER™
8012-2555
KB 65
903.00€
Each
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
30238266
View Documents
BINDER™
8012-2559
KB 130, KB 260, KB 470
957.00€
Each
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
30238246
View Documents
BINDER™
8012-2560
KB PRO 130, KB PRO 260, KB PRO 470
957.00€
Each
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
30238195
View Documents
BINDER™
8012-2561
KBF 130, KBF 260, KBF 470
957.00€
Each
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
30238198
View Documents
BINDER™
8012-2562
KB PRO 260, KB PRO 470, KB PRO 260, KB PRO 470
957.00€
Each
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
30238317
View Documents
BINDER™
8012-2563
KB 720, KB 1060
1256.00€
Each
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
30238223
View Documents
BINDER™
8012-2564
KB PRO 720, KB PRO 1060
1256.00€
Each
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
30238275
View Documents
BINDER™
8012-2565
KBF 720, KBF 1060
1256.00€
Each
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
30238334
View Documents
BINDER™
8012-2566
KBF PRO 720, KBF PRO 1060
1256.00€
Each
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
30238288
View Documents
BINDER™
8012-2568
KB PRO 1600
1608.00€
Each
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
30238202
View Documents
BINDER™
8012-2569
KBF 1600
1608.00€
Each
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
30238251
View Documents
BINDER™
8012-2570
KBF PRO 1600
1608.00€
Each
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
30238305
View Documents
BINDER™
8012-2567
KB 1600
1608.00€
Each
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
Description
Description
Interior lighting with climate-resistant LED lights, which turns on automatically when the unit's door is opened.
Specifications
Specifications
Glass
Lighting
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
BINDER™ Interior Lighting