Learn More
BINDER™ Interior Lighting
Interior lighting with climate-resistant LEDs.
Brand:  BINDER™ 8012-2562
 View more versions of this product
Product Code. 30238198
  • 957.00€ / Each
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Interior lighting with climate-resistant LED lights, which turns on automatically when the unit's door is opened.
Specifications
Specifications
KB PRO 260, KB PRO 470, KB PRO 260, KB PRO 470
Glass
Lighting
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BINDER™ Interior Lighting > For use with KB PRO 260, KB PRO 470, KB PRO 260, KB PRO 470