missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ INSTA-Blot Male Lung Tissue OncoPair
Applications: Western Blot
Brand: Novus Biologicals™ NBP2-30122
This item is not returnable.
View return policy
Description
- Proteomic discovery tool
- Addresses increasing demand for inclusion of human tissue-derived products in biomarker profiling
- Manufactured from Imgenex's extensive Human Clinical Tissue Lysate collection
- Each blot contains 14 lanes of alternating tumor (T) and matched normal adjacent (NA) tissue lysates from seven patient donors
- Incorporation of T and NA enables protein expression analysis of tumor and microenvironment
- Useful in conjunction with Cell Line, Normal Tissue INSTA-BlotsTM, lysates for comprehensive antibody validation
- Used in profile antibody reactivity, protein expression patterns across species and between tissues and cells lines
Recommended Storage
Store at room temperature

Specifications
Used in western blot for screening expression of various proteins in tumor and normal adjacent tissues from various donors | |
1 Membrane | |
PVDF |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ INSTA-Blot Male Lung Tissue OncoPair >
Spot an opportunity for improvement?Share a Content Correction