missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ INSTA-Blot Human Tissues
Applications: Western Blot
Brand: Novus Biologicals™ NBP2-31378
This item is not returnable.
View return policy
Description
Recommended Storage
Store at room temperature

Specifications
Tissues are from donors with no known disease. Tissues are lysed in lysis buffer (10 mM Tris, pH 8.0, 130 mM NaCl, 1% Triton X-100, 10 mM NaF, 10 mM NaPi, 10 mM NaPPi) containing Protease Inhibitor Cocktail (PIC) and PMSF. Prior to SDS-PAGE the tissue lysate is resuspended with 4x Laemmli sample buffer with B-mercaptoethanol and denatured in boiling water. Use in Western blot reported in scientific literature (PMID 25615412) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ INSTA-Blot Human Tissues > SDS-Page Human Tissues Membrane
Spot an opportunity for improvement?Share a Content Correction