Learn More
Elma™ Insert Covers
Insert cover contains two rubber rings and is designed for hanging cleaning cups or glasses in the cleaning basket.
Brand: Elma™ 1111909
This item is not returnable.
View return policy
Description
- Elma ultrasonic units can be operated with or without a cover
- For more efficient operation, Elma offers various insert covers made of robust and easy-to-clean stainless steel
- Covers make it possible to clean objects made of different materials simultaneously – but separately
- If used correctly, all Elma inserts will help to prevent damage to the ultrasonic tank and thus extend the service life of the ultrasonic cleaner
- Allows cleaning with aggressive fluids
- Prolongs the service life of the unit
Specifications
Lid | |
Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic S, Elmasonic Med, Elmasonic Select, Elmasonic EASY | |
100/120 |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Elma™ Insert Covers > Insert Covers; Model: 100/120