Learn More
Smeg™ Injection Trolley for Laboratory Glassware Washers
Use these trolleys with Smeg™ glassware washers. Smeg Injection Trolleys for Laboratory Glassware Washers are available in a variety of configurations to fit a wide range of applications.
Brand: Smeg™ LM80
This item is not returnable.
View return policy
Description
- For use with Smeg glassware washers
- Available with one, two and mixed spraying/injection levels
- Clean and sanitize a wide variety of labware
Specifications
Stainless Steel | |
68 items | |
Injection trolley with two injection levels | |
Laboratory glassware washers | |
1 Each |
Injection Trolley | |
two injection levels for a capacity of 68 items. | |
Washing up to 68 glassware items | |
Without drying system connection |
Product Content Correction
Product Title
Smeg™ Injection Trolley for Laboratory Glassware Washers > Model LM80; Holds: 68 items; For washing narrow-necked glassware (flasks, round-bottom flasks and graduated cylinders of various sizes)up to 225mmH