missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat/Chicken Oligodendrocyte Marker O4 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 59 publications
Brand: R&D Systems MAB1326
This item is not returnable.
View return policy
Description
Oligodendrocyte Marker O4 Monoclonal specifically detects Oligodendrocyte Marker O4 in Human, Mouse, Rat, Chicken samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
Oligodendrocyte Marker O4 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 1-10 ug/mL, CyTOF-ready | |
Mouse | |
Affinity Purified | |
RUO | |
Detects human, mouse, rat, and chicken Oligodendrocyte Marker O4. | |
Human, Mouse, Rat, Chicken | |
IgM |
Flow Cytometry, Immunocytochemistry, CyTOF | |
O4 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Bovine brain corpus callosum white matter | |
50 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Human/Mouse/Rat/Chicken Oligodendrocyte Marker O4 Antibody, R&D Systems™ > 50ug; Unlabeled
Spot an opportunity for improvement?Share a Content Correction