Learn More
Lauda™ Hydro H 20 SOW Shaking Water Bath with CEE7/7 Plug
Shaking water bath for exactly reproducible temperatures
Brand:  Lauda™ L002910
Product Code. 16659345
  • 5385.00€ / Each
Explore more special offers
Description
Show More
Specifications
Specifications
25°C to 80°C
230 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lauda™ Hydro H 20 SOW Shaking Water Bath with CEE7/7 Plug > Hydro H 20 SOW Shaking Water Bath with CEE7/7 Plug