missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human High Sensitivity Cytokine B Premixed Magnetic Luminex™ Performance Assay

Magnetic bead-based multiplex assay for the Luminex™ platform

1435.00€ - 4645.00€

Specifications

Species Human
For Use With (Application) ELISA
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
View More Specs

Products 16
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18461310
View Documents
R&D Systems™
FCSTM14-11
11 Plex
3450.00€
1 set
Please to purchase this item. Need a web account? Register with us today!
18442400
View Documents
R&D Systems™
FCSTM14-06
6 Plex
2340.00€
1 set
Please to purchase this item. Need a web account? Register with us today!
18411440
View Documents
R&D Systems™
FCSTM14-08
8 Plex
2795.00€
1 set
Please to purchase this item. Need a web account? Register with us today!
18462090
View Documents
R&D Systems™
FCSTM14-07
7 Plex
2565.00€
1 set
Please to purchase this item. Need a web account? Register with us today!
18401520
View Documents
R&D Systems™
FCSTM14-05
5 Plex
2115.00€
1 set
Please to purchase this item. Need a web account? Register with us today!
18401420
View Documents
R&D Systems™
FCSTM14-04
4 Plex
1885.00€
1 set
Please to purchase this item. Need a web account? Register with us today!
18412330
View Documents
R&D Systems™
FCSTM14-03
3 Plex
1660.00€
1 set
Please to purchase this item. Need a web account? Register with us today!
18492320
View Documents
R&D Systems™
FCSTM14-02
2 Plex
1435.00€
1 set
Please to purchase this item. Need a web account? Register with us today!
18432330
View Documents
R&D Systems™
FCSTM14-09
9 Plex
3020.00€
1 set
Please to purchase this item. Need a web account? Register with us today!
18402300
View Documents
R&D Systems™
FCSTM14-10
10 Plex
3250.00€
1 set
Please to purchase this item. Need a web account? Register with us today!
18461150
View Documents
R&D Systems™
FCSTM14-14
14 Plex
4045.00€
1 set
Please to purchase this item. Need a web account? Register with us today!
18451810
View Documents
R&D Systems™
FCSTM14-15
15 Plex
4245.00€
1 set
Please to purchase this item. Need a web account? Register with us today!
18412210
View Documents
R&D Systems™
FCSTM14-16
16 Plex
4445.00€
1 set
Please to purchase this item. Need a web account? Register with us today!
18471880
View Documents
R&D Systems™
FCSTM14-17
17 Plex
4645.00€
1 set
Please to purchase this item. Need a web account? Register with us today!
18442430
View Documents
R&D Systems™
FCSTM14-12
12 Plex
3645.00€
1 set
Please to purchase this item. Need a web account? Register with us today!
18401740
View Documents
R&D Systems™
FCSTM14-13
13 Plex
3845.00€
1 set
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Human
ELISA
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
R&D Systems™ Human High Sensitivity Cytokine B Premixed Magnetic Luminex™ Performance Assay

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.