missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human High Sensitivity Cytokine B Premixed Magnetic Luminex™ Performance Assay
Click to view available options
Quantity:
10 Plex
11 Plex
12 Plex
13 Plex
14 Plex
15 Plex
16 Plex
17 Plex
2 Plex
3 Plex
4 Plex
5 Plex
6 Plex
7 Plex
8 Plex
9 Plex
Unit Size:
1 set
Specifications
Specifications
| Quantity | 15 Plex |
| Species | Human |
| For Use With (Application) | ELISA |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction