missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human FGF-7 (KGF) Recombinant Protein, PeproTech®
Recombinant Protein
83.75€ - 5380.00€
Specifications
Accession Number | P21781 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 2252 |
Molecular Weight (g/mol) | 18.9 kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17800873
|
Gibco™
100-19-100UG |
100 μg |
1078.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17810673
|
Gibco™
100-19-10UG |
10 μg |
203.00€
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17831363
|
Gibco™
100-19-1MG |
1 mg |
5380.00€
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17850783
|
Gibco™
100-19-250UG |
250 μg |
1882.00€
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17830483
|
Gibco™
100-19-2UG |
2 μg |
83.75€
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17861523
|
Gibco™
100-19-500UG |
500 μg |
3225.00€
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17881523
|
Gibco™
100-19-50UG |
50 μg |
648.00€
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Recombinant Human KGF (FGF-7) is an 18.9 kDa protein consisting of 163 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Plays an important role in the regulation of embryonic development, cell proliferation and cell differentiation. Required for normal branching morphogenesis. Growth factor active on keratinocytes. Possible major paracrine effector of normal epithelial cell proliferation.Specifications
P21781 | |
protein with no preservative | |
18.9 kDa | |
E. coli | |
<1 EU/ μg | |
KGF | |
Determined by a cell proliferation assay using 4 MBr-5 cells. The expected ED50 for this effect is 0.1-15 ng/ml | |
Recombinant | |
-20°C | |
Lyophilized |
ELISA, Functional Assay, Western Blot | |
2252 | |
Human FGF-7 (KGF) | |
RUO | |
Fgf7; FGF-7; Fibroblast growth factor 7; HBGF-7; Heparin-binding growth factor 7; keratinocyte growth factor; Kgf | |
FGF7 | |
Unconjugated | |
CNDMTPEQMA TNVNCSSPER HTRSYDYMEG GDIRVRRLFC RTQWYLRIDK RGKVKGTQEM KNNYNIMEIR TVAVGIVAIK GVESEFYLAM NKEGKLYAKK ECNEDCNFKE LILENHYNTY ASAKWTHNGG EMFVALNQKG IPVRGKKTKK EQKTAHFLPM AIT | |
E. coli | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ Human FGF-7 (KGF) Recombinant Protein, PeproTech®