Learn More
Hemin (porcine), 97+%, Thermo Scientific Chemicals
Description
Hemin is used in cellular protection and control mechanism. It stimulates the synthesis of globulin. It is also used to study the bacterial strains and growth conditions, in the short-term colony assays and cell transfections. Further, it is used to treat the symptoms of various porphyrias. In addition, it is used in the preparation of agar medium for the bacteria culture.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Hemin is used in cellular protection and control mechanism. It stimulates the synthesis of globulin. It is also used to study the bacterial strains and growth conditions, in the short-term colony assays and cell transfections. Further, it is used to treat the symptoms of various porphyrias. In addition, it is used in the preparation of agar medium for the bacteria culture.
Solubility
Soluble in dilute ammonia, trimethylamine, p-toluidine and dimethylaniline. Slightly soluble in alcohol. Insoluble in water
Notes
Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | >300°C |
Quantity | 25 g |
Beilstein | 5229914 |
Merck Index | 14,4644 |
Solubility Information | Soluble in dilute ammonia,trimethylamine,p-toluidine and dimethylaniline. Slightly soluble in alcohol. Insoluble in water |
IUPAC Name | 3-[(1Z,6Z,12Z,17Z)-5-(2-carboxyethyl)-22-chloro-15,20-diethenyl-4,10,14,19-tetramethyl-21,23,24,25-tetraaza-22-ferrahexacyclo[9.9.3.1³,⁶.1¹³,¹⁶.0⁸,²³.0¹⁸,²¹]pentacosa-1,3(25),4,6,8,10,12,14,16(24),17,19-undecaen-9-yl]propanoic acid |
Formula Weight | 651.95 |
Percent Purity | ≥98% |
Chemical Name or Material | Hemin (porcine) |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10253457 | Certificate of Analysis | 19/12/2024 | A11165.03,A11165.14,A11165.06 |
U21K013 | Certificate of Analysis | 02/10/2024 | A11165.03,A11165.14,A11165.06 |
10251074 | Certificate of Analysis | 30/08/2024 | A11165.03,A11165.14,A11165.06 |
S14K014 | Certificate of Analysis | 28/06/2024 | A11165.03,A11165.14,A11165.06 |
Q11K010 | Certificate of Analysis | 11/06/2024 | A11165.03,A11165.14,A11165.06 |
Safety and Handling

- Hemin (porcine)
- Danger
- Respiratory sensitiser Category 1
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contains : Hemin
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.