missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Hemin, 99%, porcine Product Code.: 10286161

Thermo Scientific Chemicals Hemin, 99%, porcine

Product Code. 10286161
25 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10286161

Brand: Thermo Scientific Chemicals 345960250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hemin, 98%, porcine, CAS # 16009-13-5, is an oxidized version of heme. It is an iron-containing prosthetic group in several proteins, and can be an alternative source of iron within the host that contains a porphyrin ring containing a Fe2+ ion.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Hemin derived from porcine is an iron-containing prosthetic group present in some proteins and can be an alternative source of iron within the host that contains a porphyrin ring containing a Fe2+ ion

Applications

• Hemin is a protoporphyrin IX containing a ferric iron (Fe3+) ion with a coordinating chloride ligand

• It is used in cellular protection and controls biological mechanisms

• It can stimulate globulin synthesis

• It can be applied to study bacterial strains and growth conditions in the short-term colony assays and cell transfections

• It is used during the preparation of agar medium for bacteria culture

• This compound has been used in L929 and J774 murine macrophage cells in order to study conjugation, characterization and toxicity of lipophosphoglycan-polyacrylic acid conjugate for vaccination against leishmaniasis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16009-13-5
Molecular Formula C34H32ClFeN4O4
Molecular Weight (g/mol) 651.95
MDL Number MFCD00010726
InChI Key BTIJJDXEELBZFS-HXFTUNQESA-K
Synonym chlorohemin, hemin chloride, protoferriheme, ferriheme, ferriprotoporphyrin ix chloride, ferriprotoporphyrin ix, panhematin, protohemin, protohemin ix, ferriprotoporphyrin
PubChem CID 131675604
IUPAC Name 3-[(1Z,6Z,12Z,17Z)-5-(2-carboxyethyl)-22-chloro-15,20-diethenyl-4,10,14,19-tetramethyl-21,23,24,25-tetraaza-22-ferrahexacyclo[9.9.3.1³,⁶.1¹³,¹⁶.0⁸,²³.0¹⁸,²¹]pentacosa-1,3(25),4,6,8,10,12,14,16(24),17,19-undecaen-9-yl]propanoic acid
SMILES CC1=C(CCC(O)=O)\C2=C\C3=C(CCC(O)=O)C(C)=C4\C=C5/N=C(/C=C6\N([Fe](Cl)N34)\C(=C/C1=N2)C(C=C)=C6C)C(C=C)=C5C

Specifications

Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 4679
Formula Weight 651.95
Percent Purity ≥97.5% (HPLC)
Physical Form Liquid
Chemical Name or Material Hemin, 99%
compliance-icons
Hazard Category
  • Self-heating substance Category 1
Hazard Statement
  • H251-Self-heating: may catch fire.
Precautionary Statement
  • P235-Keep cool.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P420-Store away from other materials.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Hemin, 99%, porcine >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.