missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences Gravity flow Strep-Tactin™ 4Flow™ high capacity column
Strep-Tactin™ 4Flow™ high capacity gravity flow column to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
126.00€ - 379.00€
Specifications
Binding Capacity | 20 mg/mL resin |
---|---|
Description | 4% agarose, crosslinked |
Particle Size | 50 to 150 μm |
Form | Resin Slurry |
Protein Form | Tetrameric protein |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
30117609
|
IBA Lifesciences
2-1252-001 |
1 mL |
126.00€
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30117604
|
IBA Lifesciences
2-1253-001 |
5 mL |
379.00€
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specifications
20 mg/mL resin | |
50 to 150 μm | |
Tetrameric protein | |
Affinity chromatography equipment | |
Agarose | |
Pre-packed gravity flow column |
4% agarose, crosslinked | |
Resin Slurry | |
Proteins tagged with Twin-Strep-tag™; Strep-tag™II; cross reactivity: biotin | |
Twin-Strep-tag™; Strep-tag™II; cross reactivity: biotin | |
Gravity flow column |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IBA Lifesciences Gravity flow Strep-Tactin™ 4Flow™ high capacity column