missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Goat anti-Mouse IgA Secondary Antibody
Goat Polyclonal Secondary Antibody
Brand: Invitrogen™ SA510253
This item is not returnable.
View return policy
Description
This antibody may cross react with IgA from other species.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Mouse IgA | |
Polyclonal | |
Unconjugated | |
Goat | |
Antigen affinity chromatography | |
RUO | |
Mouse | |
Secondary Antibody | |
IgG |
ELISA, Immunoelectrophoresis, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Mouse IgA. | |
1 mg | |
Secondary | |
4°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ Goat anti-Mouse IgA Secondary Antibody > 1 mg; Unconjugated
Spot an opportunity for improvement?Share a Content Correction