missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneBLAzer™ LXRα-UAS-bla HEK 293T Cells

LXR alpha-UAS-bla HEK 293T 293 cells contain the ligand-binding domain (LBD) of the human Liver-X receptor-alpha(LXR alpha) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK 293T cell line.

Brand:  Thermo Scientific™ K1692

Product Code. 10465185

  • 10850.00€ / Each
Estimated Shipment: 02-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

LXR alpha-UAS-bla HEK 293T 293 cells contain the ligand-binding domain (LBD) of the human Liver-X receptor-alpha(LXR alpha) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK 293T cell line. GeneBLAzer™UAS-bla HEK 293T cells stably express a beta-lactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS). When an agonist binds to the LBD of the GAL4 (DBD)-LXR alpha (LBD) fusion protein, the protein binds to the UAS, resulting in expression of beta-lactamase. Division Arrested (DA) cells are available in two configurations- an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates. LXR alpha-UAS-bla HEK 293T 293 cells are functionally validated for Z' and EC50 concentrations of TO901317. In addition, LXR alpha-UAS-bla HEK 293T 293 cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cholic Acid, Deoxycholic Acid
Cell-based beta lactamase reporter gene
LXR alpha-UAS-bla HEK 293T cells (K1249)

Shipping: Dry Ice
Storage: -80°C (liquid nitrogen), Liquid Nitrogen
Antagonist: (z)-guggulsterone; Primary Agonist: Chenodeoxycholic Acid (CDCA)
Human
LXR alpha-UAS-bla HEK 293T cells (K1249)
Chenodeoxycholic Acid (CDCA)
2 x 106 Cells
BLA (Beta-Lactamase)
Dividing Cells
(z)-guggulsterone
Dividing Cells
Fluorescence, Fluorescent
Nuclear Receptors
Nuclear Receptors
2 x 106
GeneBLAzer™
HEK 293T Cells
LXRA, LXR alpha, NR1H3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1306708 Certificate TypeCertificate of Analysis Date15/04/2017 Catalog NumberK1692
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ GeneBLAzer™ LXRα-UAS-bla HEK 293T Cells > 2 x 106 Cells

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.