Learn More
Hanna Instruments™ Free and Total Chlorine High Range Reagent
For the photometric determination of free chlorine and total chlorine
Brand: Hanna Instruments™ HI93734-01
Description
- Range : 0.00 to 10.0mg/L
- Compatibility : HI-96734 portable photometer
Specifications
Free and Total Chlorine Reagent | |
For the photometric determination of free chlorine and total chlorine |