Learn More
Acetoacetyl CoA synthetase Antibody (011), PerCP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-89801PCP
Product Code. 18659360
  • 612.00€ / 0.10mL
Explore more special offers
Description
Description
Acetoacetyl CoA synthetase Monoclonal antibody specifically detects Acetoacetyl CoA synthetase in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Acetoacetyl CoA synthetase
Monoclonal
PerCP
PBS
Rabbit
Protein A purified
RUO
65985
Store at 4C in the dark.
IgG
ELISA
011
ELISA
acetoacetate-CoA ligase, acetoacetyl-CoA synthetase, ACSF1FLJ41251, Acyl-CoA synthetase family member 1, EC 6.2.1, EC 6.2.1.16, FLJ12389, homolog of C. elegans supressor of ras 5 (sur-5), Protein sur-5 homolog, SUR-5
This antibody was obtained from a rabbit immunized with purified, recombinant Human Acetoacetyl CoA synthetase (Accession#: NP_076417.2; Met1-Phe672).
0.1 mL
Primary
Human
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Acetoacetyl CoA synthetase Antibody (011), PerCP, Novus Biologicals™ > 0.1 mL; PerCP