Learn More
Brady™ Engraved Replacement Labels for M610 and M611
Engraved Plate Replacement Labels for use with Brady™ Printers M611 and M610.
Brand: Brady™ 312040
Description
- Engraved Plate Replacement labels for the M610 Portable Printer
- Aggressive adhesive and solvent resistance makes them a great replacement for engraved tags
- Service temperature of -40°C to +100°C
Specifications
M6-08-7593-WT | |
None | |
Gloss | |
Polyester | |
Thermal Transfer | |
Non-sterile | |
M610, M611, BMP61 | |
-40°C | |
Rectangle | |
-40°C to +100°C | |
0.65 mm | |
1.89 in. | |
19 mm |
Permanent Acrylic, Foam-backed | |
White | |
Label | |
Single-side Printable | |
Engraved Plate Labels | |
Data and Telecommunications, Electrical Labeling, General Identification | |
1 Label | |
300 Labels | |
-40°F to +212°F | |
0.0256 in. | |
48 x 19 mm | |
0.75 in. | |
19 mm |