Learn More
Taylor Wharton™ XT Series Long-Term Cryogenic Storage Dewar
The durable ribbed high-strength aluminium body, and superior vacuum performance and insulation provide versatile extended-time storage.
Brand: Taylor Wharton™ XTL8
This item is not returnable.
View return policy
Description
- Provides extended-time storage
- High-strength aluminium body
- Provides maximum holding times with superior vacuum performnce and insulation
Specifications
Long-term Cyrogenice Storage Dewar | |
8 L | |
Cryogenic storage |
XTL8 | |
Aluminum |
Product Content Correction
Product Title
Taylor Wharton™ XT Series Long-Term Cryogenic Storage Dewar > Capacity: 8 L