Learn More
Hirschmann™ DURAN™ Class AS Automatic Burettes White Graduations
Excellent for sensitively titrating amounts of chemical solutions in the laboratory
Brand:  Hirschmann™ 355037060
 View more versions of this product
Product Code. 10788421
  • 439.00€ / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Pellet
  • With white graduation
  • Amber glass
  • Acc. to DIN EN ISO 385
Specifications
Specifications
Brown
500 mm
0.05 mL
White
PTFE
Automatic Titrating Buret
DIN EN ISO 385
25 mL
Glass
No bottle
Lateral
±0.03 mL
Class AS
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
Hirschmann™ DURAN™ Class AS Automatic Burettes White Graduations > Capacity: 25 mL, Includes: No bottle