Learn More
Labconco™ WaterPro™ Reverse Osmosis (RO) Systems
Three filters (pre, carbon and RO membrane) combined to deliver high-quality reverse osmosis purified water
Brand: Labconco™ 9075030
Alert:
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Description
- Ideal for laboratory applications such as reagent preparation and glassware rinsing
- Obtain purified rinse water by connecting to laboratory glassware washers, such as SteamScrubber™ or FlaskScrubber™
- Timed dispense from 1 to 99.9 min. for unattended operation
- Temperature compensated conductivity LED monitor displays water quality in microsiemens from 1 to 999
- Dispense manually from valve or with dispensing gun (sold separately) at rate of 8.7L/min. (tank full) or 1L/min. (tank empty)
- Storage tank air vent filter removes bacteria, maintaining water quality
- Automatic flush valve extends RO membrane life by automatically flushing the RO membrane and tank for three minutes after each 12 hour period of inactivity
- Internal pressure gauge monitors and displays RO-purified water pressure from 0 to 160psi
- Durable Type 304 stainless steel front panel; powder-coated steel cabinet
- Large capacity white polypropylene bowls with threaded heads
- Integral 17L polyethylene storage tank
- Quiet 1/4 hp motor/rotary vane pump
- Wall mountable
Certifications
ETL listed.
Specifications
CE, ETL | |
No | |
1.1 L/min. | |
No | |
LED | |
0 to 160 psi | |
31.1 in. | |
30.8 in. | |
7.6 in. | |
90 lb. | |
230 V 50/60 Hz | |
6 A |
17 L | |
Tap | |
Type 3 to 4 | |
Reverse Osmosis System | |
Schuko | |
Carbon filtration, reverse osmosis | |
78 cm | |
78 cm | |
19.3 cm | |
40.8 kg | |
230 V | |
50/60 Hz |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.