Learn More
Thermo Scientific™ Decontamination Kit
For Use With: For Water Jacket CO2 Incubators
Brand:  Thermo Scientific™ LT2270105
Product Code. 11826385
  • 254.00€ / Each
Estimated Shipment: 18-06-2025
to see stock.
Explore more special offers
Description
For Use With: For Water Jacket CO2 Incubators
HEPA Filters
  • 99.97% filtering at 0.3μm
  • Compatible with all Thermo Scientific™ Forma™ Series II 3110, Thermo Scientific™ Steri-Cycle™, and Forma™ 310 Series (with HEPA filtration system option) incubators
  • Main chamber filter available as a stand-alone or packaged in a group with the other chamber in-line filters
Decontamination Kits
  • Consists of filters for difficult-to-clean chambers and gaskets (except door) and tubing
  • Reduces downtime and labor spent on decontamination
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Internal chamber replacement kit with HEPA, gaskets, and other parts
For Water Jacket CO2 Incubators
LED
Decontamination Kit
Product Title
Thermo Scientific™ Decontamination Kit > Decon Kit; For 3110 Series Incubators