Learn More
Thermo Scientific™ Decontamination Kit
For Use With: For Water Jacket CO2 Incubators
Brand: Thermo Scientific™ LT190651
239.40 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
This item is not returnable.
View return policy
Description
For Use With: For Water Jacket CO2 Incubators
HEPA Filters
- 99.97% filtering at 0.3μm
- Compatible with all Thermo Scientific™ Forma™ Series II 3110, Thermo Scientific™ Steri-Cycle™, and Forma™ 310 Series (with HEPA filtration system option) incubators
- Main chamber filter available as a stand-alone or packaged in a group with the other chamber in-line filters
Decontamination Kits
- Consists of filters for difficult-to-clean chambers and gaskets (except door) and tubing
- Reduces downtime and labor spent on decontamination
Specifications
Internal chamber replacement kit with HEPA, gaskets, and other parts | |
For Water Jacket CO2 Incubators | |
LED | |
Decontamination Kit |
Promotions
-
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Product Content Correction
Product Title
Thermo Scientific™ Decontamination Kit > Decon Kit; For 3110 Series Incubators