Learn More
Invitrogen™ Customer-Defined Anti-miR™ miRNA Inhibitor
Anti-miR™ miRNA Inhibitors are chemically modified, single-stranded nucleic acids designed to specifically bind to and inhibit endogenous microRNA (miRNA) molecules.
Brand:  Invitrogen™ AM17003
777.20 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Product Code. 15524394
  • 1212.00€ / 20nmol
Explore more special offers
Description
Description
Use our customization tool to design the ideal inhibitor for your miRNA sequence, or select from among predesigned products.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
The Anti-miR™ miRNA is provided dried down in a single tube, along with Nuclease-free Water for resuspension, and should be stored at -20°C. It is guaranteed for 1 year when stored dried (at -20°C.)
Ambion™, Anti-miR™
20 nmol
miRNA
miRNA Inhibitor
Product Title
Invitrogen™ Customer-Defined Anti-miR™ miRNA Inhibitor > Guaranteed for 1 year when stored dried at –20°C.