missing translation for 'onlineSavingsMsg'
Learn More

dsRNA Antibody (1D3), DyLight 405 , Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-20110V

Product Code. 18526848

  • 480.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

dsRNA Monoclonal antibody specifically detects dsRNA in Broad Species Reactivity samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

dsRNA
Monoclonal
DyLight 405
Double-stranded RNA
RDV-RNA-methylated bovine serum albumin complex.
0.1 mL
Primary
Store at 4°C in the dark.
IgG1 κ
Western Blot, ELISA, Immunohistochemistry
1D3
50mM Sodium Borate
Mouse
Protein A purified
RUO
All species
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
dsRNA Antibody (1D3), DyLight 405 , Novus Biologicals™ > 0.1 mL; DyLight 405

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.